accessibility menu, dialog, popup

UserName

CAS RN 1808990-66-0

IUPAC Name: N-{20-[3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanamido]-3,6,9,12,15,18-hexaoxaicosan-1-yl}-5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanamide
Synonyms: N-{20-[3-(2,5-dioxopyrrol-1-yl)propanamido]-3,6,9,12,15,18-hexaoxaicosan-1-yl}-5-{2-oxo-hexahydrothieno[3,4-d]imidazol-4-yl}pentanamide
Molecular Weight (g/mol): 701.83
Molecular Formula: C31H51N5O11S
InChi Key: ILVNKONXZBFCPD-UHFFFAOYNA-N
SMILES: O=C(CCCCC1SCC2NC(=O)NC12)NCCOCCOCCOCCOCCOCCOCCNC(=O)CCN1C(=O)C=CC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results