missing translation for 'onlineSavingsMsg'
Learn More

Biotin-PEG4-Azide 96.0+%, TCI America™

Catalog No. B5546100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5546100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B5546100MG Supplier TCI America Supplier No. B5546100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1309649-57-7
Molecular Formula C20H36N6O6S
Molecular Weight (g/mol) 488.604
MDL Number MFCD29495433
InChI Key PVEHVEYAPUNCCP-LNLFQRSKSA-N
Synonym N-[2-[2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]ethoxy]ethyl]biotinamide
PubChem CID 77078236
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-[2-[2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethoxy]ethyl]pentanamide
SMILES C1C2C(C(S1)CCCCC(=O)NCCOCCOCCOCCOCCN=[N+]=[N-])NC(=O)N2

Specifications

Melting Point 104°C
Color White-Yellow
Quantity 100 mg
Formula Weight 488.60
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Biotin-PEG4-Azide
TSCA No
Recommended Storage Freezer