accessibility menu, dialog, popup

UserName

CAS RN 1309649-57-7

IUPAC Name: N-(14-azido-3,6,9,12-tetraoxatetradecan-1-yl)-5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanamide
Synonyms: N-(14-azido-3,6,9,12-tetraoxatetradecan-1-yl)-5-{2-oxo-hexahydrothieno[3,4-d]imidazol-4-yl}pentanamide
Molecular Weight (g/mol): 488.6
Molecular Formula: C20H36N6O6S
InChi Key: PVEHVEYAPUNCCP-UHFFFAOYNA-N
SMILES: [N-]=[N+]=NCCOCCOCCOCCOCCNC(=O)CCCCC1SCC2NC(=O)NC12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results