missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Biotin Azide (PEG4 carboxamide-6-Azidohexanyl Biotin)
Description
The hapten, biotin azide is reactive with terminal alkynes via a copper-catalyzed click reaction. Biotin can be subsequently detected with streptavidin, avidin or NeutrAvidin™ biotin-binding protein.
Specifications
Specifications
| Format | Solid |
| Quantity | 1 mg |
| Product Type | Biotin Azide |
| Molecular Weight (g/mol) | 615.79 Da |
| Solubility | DMSO (Dimethylsulfoxide) |
| Content And Storage | Store at ≤-20°C and keep desiccated. |
| Chemical Reactivity | Alkyne |
| Reactive Group | Azide |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Amine, Azide |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.