Learn More
Applied Biosystems™ Biotin Amidite
Catalog No. 401395
Encompass
Change view
Click to view available options
Quantity:
85 mg
Catalog No. Quantity
401395 85 mg
1 options
Catalog No. 401395 Supplier Applied Biosystems™ Supplier No. 401395
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Contains 85 mg Biotin Amidite.Biotin Amidite is used for the 5’ attachment of Biotin during oligonucleotide synthesis.The label is treated as if it were simply another base: It is inserted at the appropriate place in the sequence and incorporated automatically.
On-instrument labeling typically saves hours of post-synthesis manual processing.Biotin Amidite is used to create 5'-biotinylated oligonucleotides that allow control of the oligonucleotide through the affinity of biotin for streptavidin. Capturing biotin-labeled oligonucleotides on streptavidin-coated media simplifies recovery of target sequences and immobilization for sequencing and nonisotopic assays.
  • Specialty phosphoramidite allow online addition of the label during synthesis, saving time in production of labeled probes and primers.
  • Designed and optimized for use with ABI synthesizers, guaranteeing reliable nucleic acid synthesis.
  • We offer a range of specialty phosphoramites and linkers to give you the flexibility demanded in oligonucleotide synthesis.
Specifications
Packaging Type Shipped on Dry Ice
Quantity 85 mg
For Research Use Only. Not for use in diagnostic procedures.