missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Biotin-4-Fluorescein

Catalog No. b10570
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10570 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B10570 Supplier Invitrogen™ Supplier No. B10570

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Biotin-4-fluorescein can be used to quantitate available biotin-binding sites or the concentration of avidin or streptavidin through the strong quenching associated when this molecule binds to avidin.

Biotin-4-fluorescein can be used to quantitate available biotin-binding sites or the concentration of avidin or streptavidin through the strong quenching associated when this molecule binds to avidin.

To make a concentrated stock solution of up to 1 mM, dissolve in either anhydrous DMSO, DMF or 100% methanol. Stock solutions should be stored frozen, desiccated and protected from light.

Specifications

Quantity 5 mg
Product Type Biotin-4-Fluorescein
Molecular Weight (g/mol) 644.7
Content And Storage Store at room temperature and protect from light.
Shipping Condition Room Temperature
Label or Dye FITC (Fluorescein)
Emission 523 nm
Excitation 494 nm

For Research Use Only. Not for use in diagnostic procedures.