Learn More
Agilent BioTek Washer Biomagnetic Separation Carrier Accessories
GREENER_CHOICE
Catalog No. BT7102216
Change view
Click to view available options
Type:
384-Well Ring Magnet
96-Well Ring Magnet
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
BT7102216 96-Well Ring Magnet
BT7102215 384-Well Ring Magnet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BT7102216 Supplier Agilent BioTek Supplier No. 7102216
Enables high strength biomagnetic separation
  • Magnet ring
  • External
  • For insertion into washer biomagnetic separation carriers
  • Beads pulled to 4-zone ring at well bottom
Specifications
Type 96-Well Ring Magnet
For Use With (Equipment) 405 LS Microplate Washer, 405 Touch Microplate Washer, EL406 Microplate Washer, ELx 405 Multiflo FX Microplate Washer