missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek Microplate Washer Dispense Bottles
For use with microplate washers and washer dispenser models
Supplier: Agilent BioTek 7100559
Description
- Provides supply containment
- Capacity: 10L
Specifications
| 10 L | |
| Dispense Bottle | |
| EL406 Microplate Washer, ELx 405 Microplate Washer |