Learn More
Agilent BioTek BioStack Integration Kits
GREENER_CHOICE
Catalog No. BT7310042
Change view
Click to view available options
For Use With (Equipment):
Biostack Microplate Stacker, 405 Touch Microplate Washer, 405 LS Microplate Washer
Biostack Microplate Stacker, EL406 Microplate Washer Dispenser
Biostack Microplate Stacker, ELx 405 Select Deep Well Microplate Washer
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
BT7310042 Biostack Microplate Stacker, EL406 Microplate Washer Dispenser
BT7310040 Biostack Microplate Stacker, 405 Touch Microplate Washer, 405 LS Microplate Washer
BT7310041 Biostack Microplate Stacker, ELx 405 Select Deep Well Microplate Washer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BT7310042 Supplier Agilent BioTek Supplier No. 7310042
Includes
Cat. No. BT7310042
Base for 90° and 0° integrations, four positioning feet, communication cable and interface software
Cat. No. BT7310041
Base for 90° and 0° integrations, four positioning feet and communication cable
Cat. No. BT7310040
Base for 90° and 0° integrations, four positioning feet and communication cable
Secures alignment for trouble-free performance.
  • Used with BioStack Microplate Stacker
  • Includes base for 90° and 0° integrations
  • Four positioning feet
Specifications
Includes Base for 90° and 0° integrations, 4 positioning feet and communication cable
Type Integration Kit
For Use With (Equipment) Biostack Microplate Stacker, EL406 Microplate Washer Dispenser