Learn More
Agilent BioTek BioStack Integration Kit for Synergy 2, Synergy MX, Synergy 4 and Synergy H4
For use with Synergy 2, Synergy MX, Synergy 4 and Synergy H4 microplate readers.
Supplier: Agilent BioTek 7310047
Includes: Base for 0 deg. integration, 4 positioning feet, communication cable and PC software
Description
For use with Synergy 2, Synergy MX, Synergy 4 and Synergy H4 microplate readers.Specifications
| BioStack Integration Kit | |
| Synergy 2, Synergy MX, Synergy 4 and Synergy H4 microplate readers | |
| Base for 0 deg. integration, 4 positioning feet, communication cable and PC software |