missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek BioStack Automated Microplate Stackers, IVD
GREENER_CHOICE

Catalog No. BTBSTCKN2SI
Change view
Click to view available options
Product Type:
BioStack 3 Microplate Stacker
BioStack 4 Microplate Stacker
BioStack Microplate Stacker
BioStack NEO Microplate Stacker
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
BTBSTCKN2SI BioStack NEO Microplate Stacker
BTBSTK2WRSI BioStack Microplate Stacker
BTBSTK3WRSI BioStack 3 Microplate Stacker
BTBSTCK4SI BioStack 4 Microplate Stacker
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. BTBSTCKN2SI Supplier Agilent BioTek Supplier No. BSTCKN2SI

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BioTek BioStack microplate stacker is a compact and versatile microplate stacker, compatible with Agilent BioTek washers, dispensers, plate readers, and imaging systems. For In Vitro Diagnostic Use.

BioTek BioStack microplate stacker is a compact and versatile microplate stacker, compatible with Agilent BioTek washers, dispensers, plate readers, and imaging systems. The rapid plate exchange speeds deliver increased throughput and enhanced efficiency and greater productivity

BioStack can accommodate assay workflows for 24- to 1536-well plates. Microscope slides can be stacked for imaging using BioTek's custom slide holders. The BioTek BioStack 4 is a delidding microplate stacker, automating the handling of lidded microplates for sensitive cell-based assays, for complete walkaway automation.

Specifications

Hertz 50/60 Hz
Voltage 100/240 V
Operating Temperature Range (Metric) 18 to 40°C
Certifications/Compliance CE and TUV marked. RoHS compliant.
Depth (English) 20.7 in.
Depth (Metric) 52.6 cm
Holds 10-, 30- and 50-plate stacks are removable and interchangeable; 96-/384-well plates: Up to 50 plates; 1536-well plates: Up to 75 plates
Humidity 10 to 85%
Includes One instrument integration kit; Communication cable; Operator manual; Microplate pedestal for storage stack loading and unloading; Maintenance kit
Plate Type ANSI/SBS Standard and low profile 96-, 384- and 1536-well plates, max. plate height 14.6 mm
Software LHC v 2.12 for liquid handling instruments (optional); Gen5 2.01 for readers
Weight (English) <25 lb.
Weight (Metric) 11.3 kg
Width (English) 7.4 in.
Width (Metric) 18.8 cm
Product Type BioStack NEO Microplate Stacker
Show More Show Less