missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek BioStack Automated Microplate Stackers
GREENER_CHOICE

Catalog No. BTBIOSTACK4
Change view
Click to view available options
Product Type:
BioStack 3 Microplate Stacker
BioStack 4 Microplate Stacker
BioStack Microplate Stacker
BioStack NEO Microplate Stacker
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
BTBIOSTACK4 BioStack 4 Microplate Stacker
11120601 BioStack Microplate Stacker
BIOSTACK3WR BioStack 3 Microplate Stacker
BIOSTACKNEO BioStack NEO Microplate Stacker
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. BTBIOSTACK4 Supplier Agilent BioTek Supplier No. BSTCK4SN

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

One instrument integration kit, communication cable, operator manual, microplate pedestal for storage stack loading and unloading, maintenance kit.

BioTek BioStack microplate stacker is a compact and versatile microplate stacker, compatible with Agilent BioTek washers, dispensers, plate readers, and imaging systems.

BioTek BioStack microplate stacker is a compact and versatile microplate stacker, compatible with Agilent BioTek washers, dispensers, plate readers, and imaging systems. The rapid plate exchange speeds deliver increased throughput and enhanced efficiency and greater productivity

BioStack can accommodate assay workflows for 24- to 1536-well plates. Microscope slides can be stacked for imaging using BioTek's custom slide holders. The BioTek BioStack 4 is a delidding microplate stacker, automating the handling of lidded microplates for sensitive cell-based assays, for complete walkaway automation.

Certifications

CE marked; ETL listed

Specifications

Voltage 100/240 V
Certifications/Compliance CE and TUV marked. RoHS compliant.
Depth (English) 22 in.
Depth (Metric) 56 cm
Holds 10 and 30 plate stacks are removable and interchangeable (50-plate stacks may be used with non-lidded plates only); 96-/384-well plates: Up to 30 plates (with lids); 1536-well plates: Up to 75 plates
Humidity 10 to 85%
Includes One instrument integration kit; Communication cable; Operator manual; Microplate pedestal for storage stack loading and unloading; Maintenance kit
Plate Type ANSI/SBS Standard and low profile 96-, 384- and 1536-well plates; 24- and 48-well plates (model dependent) Maximum plate height: 23.2 mm
Software LHC v 2.12 for liquid handling instruments (optional); Gen5 2.01 for readers
Weight (English) <25 lb.
Weight (Metric) 11.3 kg
Width (English) 8.3 in.
Width (Metric) 21 cm
Product Type BioStack 4 Microplate Stacker
Operating Temperature Range (Metric) 18 to 40°C
Frequency 50/60 Hz
Show More Show Less