missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek BioSpa 8 Automated Incubator, IVD
GREENER_CHOICE

Catalog No. BTBIOSPASI
Change view
Click to view available options
Includes:
1 Alignment Kit
CO2/O2 Control and Monitoring, Water Pan and Humidity Monitoring, 1 Alignment Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
BTBIOSPASI 1 Alignment Kit
BTBIOSPAGSI CO2/O2 Control and Monitoring, Water Pan and Humidity Monitoring, 1 Alignment Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTBIOSPASI Supplier Agilent BioTek Supplier No. BIOSPASI

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The BioTek BioSpa live cell analysis system offers unique benefits for a wide variety of live cell imaging applications in up to 8 microplates or other labware. For In Vitro Diagnostic Use.

The BioTek BioSpa live cell analysis system offers unique benefits for a wide variety of live cell imaging applications in up to 8 microplates or other labware. With 5 imaging modes, the system enables a wide range of microscopy applications

Capture detailed cellular images, get cell-level analyses for migration and wound healing assays, 3D spheroid formation, and many other kinetic applications. Built-in scheduling, environmental monitoring, and available liquid handling for walk away automation. Multiple users can run simultaneous processes for maximum efficiency in the lab.

Specifications

Operating Temperature Range (Metric) Ambient to 45°C
Certifications/Compliance CE and TUV marked. RoHS compliant.
Depth (English) 20.1 in.
Depth (Metric) 51 cm
Height (English) 18.9 in.
Height (Metric) 48 cm
Humidity 80 to 95%
Includes 1 Alignment Kit
Software BioSpa Software
Width (English) 27.2 in.
Width (Metric) 69 cm
Product Type Automated Microplate Incubator
Capacity 8-plate
Show More Show Less