missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek BioSpa 8 Automated Incubator, IVD
GREENER_CHOICE

The BioTek BioSpa live cell analysis system offers unique benefits for a wide variety of live cell imaging applications in up to 8 microplates or other labware. For In Vitro Diagnostic Use.

Supplier:  Agilent BioTek BIOSPAGSI

 View more versions of this product

Catalog No. BTBIOSPAGSI

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

The BioTek BioSpa live cell analysis system offers unique benefits for a wide variety of live cell imaging applications in up to 8 microplates or other labware. With 5 imaging modes, the system enables a wide range of microscopy applications

Capture detailed cellular images, get cell-level analyses for migration and wound healing assays, 3D spheroid formation, and many other kinetic applications. Built-in scheduling, environmental monitoring, and available liquid handling for walk away automation. Multiple users can run simultaneous processes for maximum efficiency in the lab.

Specifications

Chemical Identifiers

Ambient to 45°C
20.1 in.
18.9 in.
80 to 95%
27.2 in.
Automated Microplate Incubator
CE and TUV marked. RoHS compliant.
51 cm
48 cm
BioSpa Software
69 cm
8-plate

Specifications

Ambient to 45°C
20.1 in.
18.9 in.
80 to 95%
BioSpa Software
69 cm
8-plate
CE and TUV marked. RoHS compliant.
51 cm
48 cm
CO2/O2 Control and Monitoring, Water Pan and Humidity Monitoring, 1 Alignment Kit
27.2 in.
Automated Microplate Incubator
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions