missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek BioSpa 8 Automated Incubator
GREENER_CHOICE

Catalog No. 11120551
Change view
Click to view available options
Includes:
CO2/O2 Control and Monitoring, Water Pan and Humidity Monitoring, 1 Alignment Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
11120551 CO2/O2 Control and Monitoring, Water Pan and Humidity Monitoring, 1 Alignment Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11120551 Supplier Agilent BioTek Supplier No. BIOSPAGSN

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The BioTek BioSpa live cell analysis system offers unique benefits for a wide variety of live cell imaging applications in up to 8 microplates or other labware. For Research Use Only. Not for use in diagnostic procedures.

The BioTek BioSpa live cell analysis system offers unique benefits for a wide variety of live cell imaging applications in up to 8 microplates or other labware. With 5 imaging modes, the system enables a wide range of microscopy applications

Capture detailed cellular images, get cell-level analyses for migration and wound healing assays, 3D spheroid formation, and many other kinetic applications. Built-in scheduling, environmental monitoring, and available liquid handling for walk away automation. Multiple users can run simultaneous processes for maximum efficiency in the lab.

Warranty

1 year

Specifications

Operating Temperature Range (Metric) Ambient to 45°C
Certifications/Compliance CE and TUV marked. RoHS compliant.
Depth (English) 20.1 in.
Depth (Metric) 51 cm
Height (English) 18.9 in.
Height (Metric) 48 cm
Humidity 80 to 95%
Includes CO2/O2 Control and Monitoring, Water Pan and Humidity Monitoring, 1 Alignment Kit
Software BioSpa Softwate
Width (English) 27.2 in.
Width (Metric) 69 cm
Product Type Automated Microplate Incubator
Capacity 8-plate
Show More Show Less