missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek BioSpa 8 Automated Incubator

Description
The BioTek BioSpa live cell analysis system offers unique benefits for a wide variety of live cell imaging applications in up to 8 microplates or other labware. With 5 imaging modes, the system enables a wide range of microscopy applications
Capture detailed cellular images, get cell-level analyses for migration and wound healing assays, 3D spheroid formation, and many other kinetic applications. Built-in scheduling, environmental monitoring, and available liquid handling for walk away automation. Multiple users can run simultaneous processes for maximum efficiency in the lab.
Warranty
1 year
Specifications
Specifications
| Operating Temperature Range (Metric) | Ambient to 45°C |
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| Depth (English) | 20.1 in. |
| Depth (Metric) | 51 cm |
| Height (English) | 18.9 in. |
| Height (Metric) | 48 cm |
| Humidity | 80 to 95% |
| Includes | 1 Alignment Kit |
| Software | BioSpa Softwate |
| Width (English) | 27.2 in. |
| Show More |