missing translation for 'onlineSavingsMsg'
Learn More

Biomedical Polymers Translucent Multipurpose Containers

Catalog No. 22544230
Change view
Click to view available options
Capacity (Metric):
237 mL
473 mL
946 mL
1900 mL
2500 mL
5100 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
22544230 5100 mL
22544225 237 mL
22544226 473 mL
22544227 946 mL
22544228 1900 mL
22544229 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22544230 Supplier Biomedical Polymers Supplier No. BMPCN814

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-quality, non-graduated stackable containers with tight-fitting lids

  • Ideal for storing biologically hazardous samples for terminal autoclaving before disposal
  • Molded from either high-density polyethylene or polypropylene, containers are inert to most chemicals
  • Resistant to freezing and boiling
  • Easy to write on with either pen or pencil

Specifications

Capacity 172.4 oz., 5100 mL
Capacity (Metric) 5100 mL
Color Translucent
Material Polyethylene
Closure Color Translucent
Capacity (English) 172.4 oz.