Learn More
Biomedical Polymers Translucent Multipurpose Containers
High-quality, non-graduated stackable containers with tight-fitting lids
Supplier: Biomedical Polymers BMPCN812
Description
- Ideal for storing biologically hazardous samples for terminal autoclaving before disposal
- Molded from either high-density polyethylene or polypropylene, containers are inert to most chemicals
- Resistant to freezing and boiling
- Easy to write on with either pen or pencil
Chemical Identifiers
| Translucent | |
| Translucent |
Specifications
| 86 oz., 2500mL | |
| 86 oz. | |
| Polyethylene |
| 2500 mL | |
| Translucent | |
| Translucent |