Learn More
Justrite™ Biohazard Waste Cans with Foot-operated Self-closing Cover
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ Biohazard Waste Cans with Foot-Operated Self-Closing Cover provides a safe, temporary solution for collecting biological waste. This self-closing model meets NFPA1 and the International Fire Code. It shuts and locks automatically when a fusible link melts at 165°F (74°C) under fire conditions.
$231.48 - $319.90
Specifications
Certifications/Compliance OSHA; FM and TUV Approved
Color Red
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (English) Closure Type Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Closure Type Price Quantity & Availability  
19822422
View Documents Promotion Details
Justrite™
05910R
6 gal. Auto-close
Each for $231.48
Only null left
19041196
View Documents Promotion Details
Justrite™
05930R
10 gal. Self-close
Each for $319.90
Only null left
Description
Description
  • Protect workers, Reduce fire risks, and improve productivity by storing flammable fuels and chemicals in code-compliant safety cabinets
  • Designed to meet OSHA and NFPA standards, Sure-Grip EX cabinets are constructed of sturdy 18-gauge thick double-wall welded steel with 1-1/2 in. of insulating air space for fire resistance
  • High performance, self-latching doors close easily and securely for maximum protection under fire conditions
  • Fail-safe closing mechanism ensures three-point stainless steel bullet latching system works every time
  • For added security, U-Loc handle comes with a cylinder lock and key set or accepts an optional padlock
  • Haz-Alert reflective warning labels are highly visible under fire conditions or during power outages when illuminated with a flashlight
  • Patented SpillSlope galvanized steel shelves direct spills to back and bottom of a leakproof bottom sump
  • They adjust on 3 in. centers for versatile storage
  • Cabinets also feature dual vents with flame arresters, four adjustable self-leveling feet, grounding connector, and trilingual warning label
  • Durable lead-free epoxy/polyester powder-coat finish provides excellent chemical resistance
  • Additionally, acid/corrosive safety cabinets include polyethylene trays attached to galvanized steel shelves and a separate polyethylene liner for the bottom sump to resist aggressive chemicals
  • Easily remove the liner for quick cleaning of drips and leaks
  • An all epoxy baked-on powder-coat finish, inside and out, provides increased chemical resistance
  • Biohazard bags sold separately (No. 05901)
Specifications
Specifications
OSHA; FM and TUV Approved
Red