Learn More
Brady™ Biohazard Labels Vinyl BIOHAZARD
Identify the actual or potential presence of a biologically hazardous material using Biohazard Labels made from self-sticking vinyl (B-585) material.
Supplier:  Brady™ 20334LSA
 View more versions of this product
Catalog No. 17108801
Only null left
Description
Description
  • Good resistance to most common chemicals, oils and water
  • Warning labels support compliance with OSHA 29 CFR 1910.1030 Regulation for Bloodborne Pathogens Labeling
  • Warning labels support compliance with DOT 49 CFR 172.400(b) Labeling Requirements for transportation
  • Service temperature of -40°F to +122°F (-40°C to +50°C)
Specifications
Chemical Identifiers
Vinyl
Specifications
Vinyl
20 Pk.
3.5 x 5 in.
Product Suggestions
Product Suggestions