Learn More
Brady™ Biohazard Labels Vinyl BIOHAZARD
Catalog No. p-200136750
Change view
Click to view available options
Dimensions (L x W):
108 ft. x 2 in.
2.875 x 2 in.
3.5 x 5 in.
5 x 3.5 in.
10 x 7 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
17108798 2.875 x 2 in.
17108801 3.5 x 5 in.
17108799 5 x 3.5 in.
17108804 10 x 7 in.
17108809 108 ft. x 2 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17108798 Supplier Brady™ Supplier No. 18765LSA
Only null left
Identify the actual or potential presence of a biologically hazardous material using Biohazard Labels made from self-sticking vinyl (B-585) material.
  • Good resistance to most common chemicals, oils and water
  • Warning labels support compliance with OSHA 29 CFR 1910.1030 Regulation for Bloodborne Pathogens Labeling
  • Warning labels support compliance with DOT 49 CFR 172.400(b) Labeling Requirements for transportation
  • Service temperature of -40°F to +122°F (-40°C to +50°C)
Specifications
Material Vinyl
No. per Pack 25 Pk.
Dimensions (L x W) 2.875 x 2 in.