missing translation for 'onlineSavingsMsg'
Learn More

BIOTIX™ Disposable 50mL Reagent Reservoirs

Catalog No. 12111097
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Packaging:
1/Bag
5/Bag
Bulk
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Packaging
12111097 Non-sterile Bulk
12111096 Sterile 1/Bag
12111095 Sterile 5/Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12111097 Supplier BIOTIX™ Supplier No. 63300132

Easy-to-handle pressure-formed reservoir for use with automated platforms.

  • Made from polystyrene
  • With high-accuracy pour spouts
  • Chemical resistant
  • With graduation marks

Specifications

Color White
Capacity (Metric) 50 mL
Material Polystyrene
Sterility Non-sterile
Type Reservoir
For Use With (Application) Filling Multi-channel Pipettors
Packaging Bulk
Quantity 100/Cs.
Autoclavable Autoclavable
Channels 12
Disposable Single-use
Bottom Shape Flat
Show More Show Less