missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Biocytin TMR (5-(and-6)-Tetramethylrhodamine Biocytin)

Catalog No. T12921
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12921 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T12921 Supplier Invitrogen™ Supplier No. T12921

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Biocytin TMR (5-(and-6)-Tetramethylrhodamine Biocytin)

The water-soluble neuronal tracer 5-(and-6)-tetramethylrhodamine biocytin (biocytin TMR) can be detected in fixed tissues by using avidin or streptavidin probes. However, tetramethylrhodamine biocytin can also be observed directly in fixed or unfixed samples because the attached tetramethylrhodamine dye produces bright, orange-fluorescence (absorption/emission maxima ∼554/581 nm).

Specifications

Conjugate Classic Dyes
Quantity 5 mg
Content And Storage Store at room temperature and protect from light.
Shipping Condition Room Temperature
Form Solid
Product Line Invitrogen
Product Type Biocytins

For Research Use Only. Not for use in diagnostic procedures.