missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Fibrillar Collagen 24-well Multiwell Insert System with 3 μm PET Membrane

Catalog No. 15100202
Change view
Click to view available options
Diameter (Metric):
6.4 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
15100202 6.4 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15100202 Supplier Corning™ Supplier No. 354803
Only null left

Ready to use

Automation-friendly design.
  • Produces a differentiated monolayer of Caco-2 cells in vitro in just three days
  • Fibrillar collagen-coated inserts with specialized BD BioCoat media speed cell differentiation
  • Large-volume feeder tray reduces the need for frequent media changes
  • Lot-to-lot testing ensures consistency
  • Material [Membrane]: Polyethylene Terephthalate
  • Sterile
  • Pore Size: 3 μm
  • Surface Coating: Fibrillar Collagen
  • Well Count: 24
  • Surface Area [Metric]: 0.3cm2
  • Includes: 24 multiwell insert plate, specialised media to perform 24 assays, feeder tray and lid
  • Format: 24-wellClosure: Yes

Specifications

Closure Included Yes
Description Surface area: 0.3 cm2
Color Clear
Culture Area 0.2 cm2
Sterility Sterile
Surface Treatment Fibrillar Collagen
Type Multi-well Insert System
Well Count 24
For Use With (Equipment) Falcon™ companion plate
Includes 24 multiwell insert plate, specialised media to perfom 24 assays, feeder tray and lid
Pore Size 3μm
Storage Requirements 2°C to 8°C away from direct sunlight. Do not freeze
Supportive Membrane 3μm PET membrane
Diameter (Metric) 6.4 mm
Show More Show Less