Learn More
Corning™ BioCoat™ Collagen IV TC-Treated Culture Dishes
GREENER_CHOICE
Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
$582.83
Specifications
Bottom Shape Round
Material Polystyrene
Shape Round
Surface Treatment Collagen Type IV
Type Cell Culture Dish
View More Specs
Products 1
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
0877432
GREENER_CHOICE
View Documents Promotion Details
Corning™
354416
60 mm
Case of 20 Each for $582.83
Only null left
Description
Description
Microporous membranes permit diffusion of ions, low-molecular-weight proteins and other substances to both apical and basolateral cell surfaces, simulating the natural environment of cells in vivo.
  • Source: Mouse
  • TC-treated
  • Nonsterile
For adhesion, growth and differentiation of neuronal and transfected cell lines
Specifications
Specifications
Round
Round
Cell Culture Dish
Polystyrene
Collagen Type IV