Learn More
Ansell BioClean™ Synergy Non-Sterile Nitrile Gloves
Catalog No. 19501130
Change view
Click to view available options
Size:
Small
Medium
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19501130 Medium
17609998 Small
17609999 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning: No glove provides complete protection against all chemicals. Users must test the gloves against the particular chemicals and environment in which they will be used.
Catalog No. 19501130 Supplier Ansell Supplier No. BSANS
Only null left
Warning: No glove provides complete protection against all chemicals. Users must test the gloves against the particular chemicals and environment in which they will be used.
Non-Sterile nitrile gloves offer resistance to a range of chemicals, feature easy double-donning, and have low particle count.
  • Resistant to a range of chemicals
  • Powder, sulphur and accelerator free
  • Beaded cuff
  • Easy double-donning
  • Non-particulating EasyTear packaging
  • Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications
  • Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425
  • Processed in an NEBB certified ISO Class 4 environment
  • Complies with the requirements of European Glove Standards EN420:2003 +A1:2009, EN374-1:2016, EN374-5:2016 and EN421:2010
  • Physical properties comply with European Medical Glove Standard EN455-2:2015
  • Suitable for contact with dry, alcoholic and acidic foodstuffs. Regulation (EC)1935/2004 and (EC) 2023/2006
Specifications
Accelerator Free Yes
Color Green
Latex Free No
Material Nitrile
Size Medium
Sterility Non-sterile
Quantity 100 pieces per sealed inner PE bag; one inner PE bag per sealed outer PE bag; 10 outer bags per lined carton (1000 pieces)
Length (English) 12 in.
Length (Metric) 30 cm
Five (5) years from date of manufacture. Store in a dry, cool place (≤40°C) away from direct sunlight and fluorescent light.