missing translation for 'onlineSavingsMsg'
Learn More

Ansell BioClean-C Chemotherapy Protective Sleeve Covers

BioClean-C sleeve covers provide protection against a wide range of cytotoxic drugs. Manufactured from lightweight and low-linting CleanTough™ material.

$543.97 - $543.97

Specifications

Construction Ultrasonically bonded seams covered with protective tape
Product Type Controlled Environment Sleeve
Size Standard
Material Microporous Fabric
Color Blue
View More Specs

Products 1
Catalog Number Mfr. No. Sterility Price Quantity  
Catalog Number Mfr. No. Sterility Price Quantity  
19-668-257
View Documents
Ansell
SBCSC
Sterile
Case of 180 Each for $543.97
Only null left
 
Description

Description

  • Elasticated for secure fit
  • Lightweight low-linting CleanTough material
  • Silicone-free
  • Certifications and testing:
    • Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications
    • Conforms to Category 3 - Personal Protective Equipment Regulation (EU) 2016/425
    • Tested against standard EN ISO 13034:2005 + A1:2009 protective clothing against liquid chemicals (Type PB [6])
    • Tested against standard EN 14325 protective clothing against chemicals
    • Tested against permeation standard ASTM F739-12
    • Processed in an NEBB certified ISO Class 4 environment
    • Tested against standards: EN 530, Abrasion resistance; EN 6530,
    • Resistance to penetration by liquids; EN 13934-1, Tensile strength; EN 9073-4, trapezoidal tear resistance; EN 863, Puncture resistance; and EN 13935-2, Seam strength.
Specifications

Specifications

Ultrasonically bonded seams covered with protective tape
Standard
Blue
Ultrasonically Bonded with Protective Tape
BCSC
BioClean-C™ Sterile Chemotherapy Protective Aprons
EN ISO 13034:2005 + A1:2009, EN 14325, EN 530, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2
Controlled Environment Sleeve
Microporous Fabric
Elastic Ends
Elastic
Cleanroom, Clinical, Medical, Laboratory, Pharmaceutical
ASTM D6978-05, ASTM F739-12
Class 10 (ISO 4)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'