missing translation for 'onlineSavingsMsg'
Learn More

Ansell BioClean-C Chemotherapy Protective Sleeve Covers

Catalog No. 19668257
Change view
Click to view available options
Sterility:
Sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
19668257 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19668257 Supplier Ansell Supplier No. SBCSC
Only null left

BioClean-C sleeve covers provide protection against a wide range of cytotoxic drugs. Manufactured from lightweight and low-linting CleanTough™ material.

  • Elasticated for secure fit
  • Lightweight low-linting CleanTough material
  • Silicone-free
  • Certifications and testing:
    • Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications
    • Conforms to Category 3 - Personal Protective Equipment Regulation (EU) 2016/425
    • Tested against standard EN ISO 13034:2005 + A1:2009 protective clothing against liquid chemicals (Type PB [6])
    • Tested against standard EN 14325 protective clothing against chemicals
    • Tested against permeation standard ASTM F739-12
    • Processed in an NEBB certified ISO Class 4 environment
    • Tested against standards: EN 530, Abrasion resistance; EN 6530,
    • Resistance to penetration by liquids; EN 13934-1, Tensile strength; EN 9073-4, trapezoidal tear resistance; EN 863, Puncture resistance; and EN 13935-2, Seam strength.

Specifications

Product Type Controlled Environment Sleeve
Size Standard
Construction Ultrasonically bonded seams covered with protective tape
Material Microporous Fabric
Color Blue
Sterility Sterile
Closure Type Elastic Ends
Seam Type Ultrasonically Bonded with Protective Tape
Cuff Style Elastic
Model BCSC
For Use With (Application) Cleanroom, Clinical, Medical, Laboratory, Pharmaceutical
For Use With (Equipment) BioClean-C™ Sterile Chemotherapy Protective Aprons
Certifications/Compliance ASTM D6978-05, ASTM F739-12
Certifications/Compliance (Europe) EN ISO 13034:2005 + A1:2009, EN 14325, EN 530, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2
Cleanroom Class Class 10 (ISO 4)
Show More Show Less