Learn More
Agilent BioTek BioCell Vessel and Adapter
For use with: Epoch, Synergy H1, Synergy HTX.
Supplier: Agilent BioTek 7270512
Description
BioCell Adapter Plate Assembly can hold up to 8 BioCell cuvets (BT7272051) for 1 cm pathlength measurements in a Agilent BioTek microplate spectrophotometer.Chemical Identifiers
| Epoch, Synergy H1, Synergy HTX |
Specifications
| Adapter Plate Assembly | |
| Epoch, Synergy H1, Synergy HTX | |
| 1cm pathlength measurements |