Learn More
BioAcryl-P (30%, 19:1)
Premixed solution for preparing protein PAGE gels
$222.84 - $412.60
Specifications
| Chemical Name or Material | Acrylamide/Bisacrylamide 19:1 |
|---|---|
| Physical Form | Liquid |
| Concentration | 30% soln |
| Color | Colorless |
| MDL Number | MFCD00080848 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ60126AP
|
N/A | 500 mL |
Each for $222.84
|
|
|||||
|
AAJ60126K2
|
N/A | 1 L |
Each for $412.60
|
|
|||||
Description
Premixed solution for preparing protein PAGE gels
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Acrylamide/Bisacrylamide 19:1 | |
| 30% soln | |
| MFCD00080848 |
| Liquid | |
| Colorless | |
| UN3426 |
Safety and Handling
GHS H Statement
H340-H350-H372-H361-H302-H315-H319-H317
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P362-P501c
H302+H332-H315-H317-H319-H340-H350-H361f-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ACRYLAMIDE SOLUTION
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only