missing translation for 'onlineSavingsMsg'
Learn More

BioAcryl-P (30%, 19:1)

Catalog No. AAJ60126AP
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60126AP 500 mL
AAJ60126K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60126AP Supplier Thermo Scientific Chemicals Supplier No. J60126AP

Premixed solution for preparing protein PAGE gels

Premixed solution for preparing protein PAGE gels

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Acrylamide/Bisacrylamide 19:1
Physical Form Liquid
Concentration 30% soln
Color Colorless
Quantity 500 mL
MDL Number MFCD00080848
UN Number UN3426

RUO – Research Use Only