missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Synergy LX Multi-Mode Reader

The BioTek Synergy LX multi-mode reader automates many common microplate assays. NOTE: Fluorescence filter cubes are required for fluorescence measurements (sold separately). For Research Use Only. Not for use in diagnostic procedures.

Supplier:  Agilent BioTek SLXFTSSN

 View more versions of this product

Catalog No. BTSLXFTS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

The BioTek Synergy LX multi-mode reader automates many common microplate assays. The high-quality optic design ensures excellent data in absorbance, fluorescence, and luminescence detection modes. This affordable system is ideal for many common assays including nucleic acid and protein quantification, ELISA, BCA, Bradford, and cell viability.

The data display, output to a USB flash drive, printer, or Gen5 software make Synergy LX a versatile assay workstation. The large touch screen user interface and onboard software simplify programming and save time in operation of microplate and microvolume assays.

NOTE: Fluorescence filter cubes are required for fluorescence measurements (sold separately).

Certifications

CE and TUV marked. RoHS compliant.

Warranty and Services

12 Months

Specifications

Specifications

200 to 900 nm
CE and TUV marked. RoHS compliant.
38.1 cm
24 sec. (96 wells), 76 sec. (384 wells)
Monochromator Bandpass
15 in.
38.1 cm
Reader with Touchscreen
SLXFTS
24 VDC power supply compatible with 100/240 VAC at 50/60 Hz, 60 W (max.)
Fluorescence Intensity: Fluorescein 2 pM; Luminesence: 10 amol ATP
English
100/240 V
12.3 kg
38.1 cm
Xenon Flash
Wavelength: ±2 nm; Optical Density: <1% at 2.0 OD, <3% at 2.5 OD
≤5 nm
15 in.
Linear, Orbital, Double Orbital
Nucleic acid and protein quantification, ELISA, BCA, Bradford and cell viability
50/60 Hz
<1% from 0 to 2.5 OD
Fluorescence, Luminescence
Multi-mode Microplate Reader
0.001 OD
2
1 year
27 lb.
15 in.
Photodiode, PMT
End Point, Kinetic, Area Scanning, Absorbance Spectral Scanning
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions