missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Synergy LX Multi-Mode Reader
GREENER_CHOICE

Catalog No. BTSLXA
Change view
Click to view available options
Measurement Technology:
Absorbance
Absorbance, Fluorescence, Luminescence
Fluorescence, Luminescence
Includes:
Reader Only
Reader with Touchscreen
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Technology Includes
BTSLXA Absorbance Reader Only
BTSLXATS Absorbance Reader with Touchscreen
BTSLXF Fluorescence, Luminescence Reader Only
BTSLXFTS Fluorescence, Luminescence Reader with Touchscreen
BTSLXFA Absorbance, Fluorescence, Luminescence Reader Only
BTSLXFATS Absorbance, Fluorescence, Luminescence Reader with Touchscreen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. BTSLXA Supplier Agilent BioTek Supplier No. SLXASN

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The BioTek Synergy LX multi-mode reader automates many common microplate assays. NOTE: Fluorescence filter cubes are required for fluorescence measurements (sold separately). For Research Use Only. Not for use in diagnostic procedures.

The BioTek Synergy LX multi-mode reader automates many common microplate assays. The high-quality optic design ensures excellent data in absorbance, fluorescence, and luminescence detection modes. This affordable system is ideal for many common assays including nucleic acid and protein quantification, ELISA, BCA, Bradford, and cell viability.

The data display, output to a USB flash drive, printer, or Gen5 software make Synergy LX a versatile assay workstation. The large touch screen user interface and onboard software simplify programming and save time in operation of microplate and microvolume assays.

NOTE: Fluorescence filter cubes are required for fluorescence measurements (sold separately).

Certifications

CE and TUV marked. RoHS compliant.

Warranty

12 Months

Specifications

Absorbance Range 200 to 900 nm
Accuracy Wavelength: ±2 nm; Optical Density: <1% at 2.0 OD, <3% at 2.5 OD
Certifications/Compliance CE and TUV marked. RoHS compliant.
Bandwidth ≤5 nm
Depth (Metric) 38.1 cm
Depth (English) 15 in.
Measurement Speed 24 sec. (96 wells), 76 sec. (384 wells)
Shaking Linear, Orbital, Double Orbital
Wavelength Selection Monochromator
Filters Monochromator Bandpass
For Use With (Application) Nucleic acid and protein quantification, ELISA, BCA, Bradford and cell viability
Height (English) 12 in.
Frequency 50/60 Hz
Height (Metric) 30.5 cm
Linearity <1% from 0 to 2.5 OD
Includes Reader Only
Measurement Technology Absorbance
Model SLXA
Product Type Multi-mode Microplate Reader
Power Consumption 24 VDC power supply compatible with 100/240 VAC at 50/60 Hz, 60 W (max.)
Resolution 0.001 OD
Sensitivity Fluorescence Intensity: Fluorescein 2 pM; Luminesence: 10 amol ATP
USB Connections 2
Software Language Versions English
Warranty 1 year
Voltage 100/240 V
Weight (English) 27 lb.
Weight (Metric) 12.3 kg
Width (English) 15 in.
Width (Metric) 38.1 cm
Detector Type Photodiode, PMT
Software Type Gen5 Data Analysis Software
Light Source Xenon Flash
Reading Options Endpoint, Kinetic, Area Scanning, Absorbance Spectral Scanning
Show More Show Less