missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bio-Serv™ PRANG™, Oral Rehydrator Powder
Excellent for crashing, recovering and debilitation conditions
$205.33 - $205.69
Specifications
| Product Type | Oral Rehydrator |
|---|---|
| Calories | 19.88 |
| Components | Electrolyte, Vitamin C, Mineral, Dextrose, Sodium Chloride, Sodium Bicarbonate, Citric Acid, Potassium Chloride, Ascorbic Acid, Acesulfame Potassium, Natural and Artificial Flavors, Food Dye |
| Format | Powder |
| For Use With (Application) | Crashing, recovering, debilitating |
| Catalog Number | Mfr. No. | Quantity | Flavor | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Flavor | Price | Quantity | ||||
14726438
|
Bio-Serv™
F2351B |
1000 g | Banana |
Each for $205.33
|
|
||||
14-726-441
|
Bio-Serv™
F2351O |
1000 g | Orange |
Each for $205.33
|
|
||||
14726442
|
Bio-Serv™
F2351S |
1000 g | Strawberry |
Each for $205.69
|
|
||||
Description
- Flavored oral rehydration with electrolytes and Vitamin C
- Balanced mineral and glucose levels for quick energy
- Provides the benefit of electrolyte drinks with less sugar
- Powder form, just add water
- Nutritionally assayed
- Certified (contaminant screened)
Ingredients: Dextrose, Sodium Chloride, Sodium Bicarbonate, Citric Acid, Potassium Chloride, Ascorbic Acid, Acesulfame Potassium, Natural and Artificial Flavors, Food Dye
Specifications
| Oral Rehydrator | |
| Electrolyte, Vitamin C, Mineral, Dextrose, Sodium Chloride, Sodium Bicarbonate, Citric Acid, Potassium Chloride, Ascorbic Acid, Acesulfame Potassium, Natural and Artificial Flavors, Food Dye | |
| Crashing, recovering, debilitating | |
| Cool dry place |
| 19.88 | |
| Powder | |
| Powder |