Learn More
Bio-Serv™ PRANG™, Oral Rehydrator Powder
Excellent for crashing, recovering and debilitation conditions
Supplier: Bio-Serv™ F2351B
Description
- Flavored oral rehydration with electrolytes and Vitamin C
- Balanced mineral and glucose levels for quick energy
- Provides the benefit of electrolyte drinks with less sugar
- Powder form, just add water
- Nutritionally assayed
- Certified (contaminant screened)
Ingredients: Dextrose, Sodium Chloride, Sodium Bicarbonate, Citric Acid, Potassium Chloride, Ascorbic Acid, Acesulfame Potassium, Natural and Artificial Flavors, Food Dye
Specifications
| Oral Rehydrator | |
| Electrolyte, Vitamin C, Mineral, Dextrose, Sodium Chloride, Sodium Bicarbonate, Citric Acid, Potassium Chloride, Ascorbic Acid, Acesulfame Potassium, Natural and Artificial Flavors, Food Dye | |
| Powder | |
| Powder | |
| Cool dry place |
| 19.88 | |
| Banana | |
| Crashing, recovering, debilitating | |
| 1000 g |