missing translation for 'onlineSavingsMsg'
Learn More

Bio-Serv™ InnoDome™ and InnoWheel™ Enrichment Device, Certified

InnoDome and InnoWheel devices increase activity and reduce fighting when used together

$1,461.36 - $1,616.82

Specifications

Autoclavable Not Autoclavable
Color Red
Material PETE
Disposable Single-use
For Use With (Application) Addition to breeding programs
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
14726578
Encompass
View Documents Promotion Details
Bio-Serv™
S3175
InnoWheel
Case of 150 Each for $1,461.36
Only null left
 
14726577
Encompass
View Documents Promotion Details
Bio-Serv™
S3174
InnoDome
Case of 300 Each for $1,616.82
Only null left
 
Description

Description

  • 100% PETE—disposable, recyclable plastic
  • Very low profile design fits in most mouse cage systems
  • InnoDome and InnoWheel devices together are 3.027 in.H (7.69cm)
  • Transparent red color provides ease of visibility
  • Sterile

These products are not designed to be put through a cagewasher or autoclave.

Specifications

Specifications

Not Autoclavable
PETE
Addition to breeding programs
Red
Single-use
Sterile
Videos
SDS
Documents

Documents