missing translation for 'onlineSavingsMsg'
Learn More

Bio-Serv™ Crawl Balls™ Enrichment Device, Certified

Provides a comfortable hiding place for rodents

Supplier:  Bio-Serv™ K3329

Encompass

 View more versions of this product

Catalog No. 360993895


Only null left

Description

Description

  • 4 in. (10.2cm) dia. with three 2.5 in. (6.4cm) openings
  • Made of high-temperature polycarbonate
  • Transparent for ease of visibility
  • Autoclavable
  • Cagewasher safe
  • Certified (Contaminant screened)
Specifications

Chemical Identifiers

Crawl Ball
Polycarbonate
10.16 cm
Autoclavable
4 in.

Specifications

Autoclavable
Red
4 in.
Crawl Ball
Polycarbonate
10.16 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions