Learn More
Bio Plas™ Hexa-Flex™ Safety Caps
Used to cap and recap blood collection and disposable glass culture tubes
$152.82 - $152.86
Specifications
For Use With (Equipment) 10, 12, 13, 16 and 18 mm Blood Collection and Culture Tubes
View More Specs
Products 7
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
22-228-480
View Documents
N/A Natural
Pack of 500 Each for $152.82
Case of 10 Pack for $1,528.20
Only null left
22-228-481
View Documents
N/A Red
Pack of 500 Each for $152.86
Case of 10 Pack for $1,528.60
Only null left
22-228-482
View Documents
N/A Lavender
Pack of 500 Each for $152.86
Case of 10 Pack for $1,528.60
Only null left
22-228-483
View Documents
N/A Green
Pack of 500 Each for $152.82
Case of 10 Pack for $1,528.20
Only null left
22-228-485
View Documents
N/A Gray
Pack of 500 Each for $152.82
Case of 10 Pack for $1,528.20
Only null left
22-228-486
View Documents
N/A Yellow
Pack of 500 Each for $152.82
Case of 10 Pack for $1,528.20
Only null left
22-228-484
View Documents
N/A Blue
Pack of 500 Each for $152.82
Case of 10 Pack for $1,528.20
Only null left
Description
Description
Used to cap and recap blood collection and disposable glass culture tubes
  • Liquid-tight seal ensures sample integrity
  • Prevents cross-contamination and protects against spillage
  • Assorted colors available for sample identification
  • Fits six different sized tubes
Biomedical and industrial
Specifications
Specifications
10, 12, 13, 16 and 18 mm Blood Collection and Culture Tubes
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'