Learn More
Promega Bio-Glo™ Luciferase Assay System
Description
- Stable, Inhibitor-Resistant Luciferase Reagent for Bioassays
- Brighter, longer-lasting signal enables long incubations
- Room-temperature storage provides added convenience
Spécifications
Spécifications
| Quantity | 10 mL |
| Content And Storage | Varies Among Components |