Learn More
Innovatek BinaxNow Rapid Test Kits
Innovatek BinaxNow Rapid Test Kits provide results within 15 minutes
$763.63
Specifications
Reaction Speed 15 min.
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Disease Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Disease Price Quantity & Availability  
361045559
View Documents Promotion Details
Innovatek
4010WL416000
22 Tests/Kit Influenza A and B
Pack of 22 Each for $763.63
Only null left
Description
Description
  • Influenza A & B Test Kit
    • Rapid detection of Influenza A and B Nucleoprotein Antigens
    • Used for nasopharyngeal (NP) swab, nasal swab samples, and nasal wash/aspirate specimens
    • Results in 15 minutes
  • Malaria Test Kit
    • Rapid detection of Plasmodium antigens circulating in human venous and capillary EDTA whole blood
    • Differential diagnosis of Plasmodium falciparum (P.f.) infections from other less virulent malarial infections
    • Results in 15 minutes
Specifications
Specifications
15 min.