missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BIN1 Recombinant Rabbit Monoclonal Antibody (23GB3010)

Catalog No. MA553343
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553343 20 μL
MA553342 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553343 Supplier Invitrogen™ Supplier No. MA553343
Only null left

Rabbit Recombinant Monoclonal Antibody

BIN1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene encodes several isoforms of a nucleocytoplasmic adaptor protein, one of which was initially identified as a MYC-interacting protein with features of a tumor suppressor. Isoforms that are expressed in the central nervous system may be involved in synaptic vesicle endocytosis and may interact with dynamin, synaptojanin, endophilin, and clathrin. Isoforms that are expressed in muscle and ubiquitously expressed isoforms localize to the cytoplasm and nucleus and activate a caspase-independent apoptotic process. Studies in mouse suggest that this gene plays an important role in cardiac muscle development. Alternate splicing of the gene results in ten transcript variants encoding different isoforms. Aberrant splice variants expressed in tumor cell lines have also been described.

Specifications

Antigen BIN1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3010
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BIN1
Gene Accession No. O00499, O08539, O08839
Gene Alias ALP-1; AMPH 2; Amph2; amphiphysin 2; amphiphysin II; amphiphysin IIamph2; amphiphysin-like; amphiphysin-like protein; AMPHL; Bin1; box dependant MYC interacting protein 1; box-dependent myc-interacting protein 1; BRAMP-2; bridging integrator 1; DKFZp547F068; MGC10367; MGC105358; myc box dependent interacting protein 1; myc box-dependent-interacting protein 1; SH3 domain-containing protein 9; SH3P9; similar to bridging integrator 1 isoform 9
Gene Symbols BIN1
Host Species Rabbit
Immunogen A synthesized peptide derived from human BIN1 (400-593AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117028, 274, 30948
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less