missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BIN1 Monoclonal Antibody (K70035_11G6), Invitrogen™
Mouse Monoclonal Antibody
$1,557.73 - $2,745.56
Specifications
| Antigen | BIN1 |
|---|---|
| Clone | K70035_11G6 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: AGKIASNVQK KLTRAQEKVL QKLGKADETK DEQFEQCVQN FNKQLTEGTR LQKDLRTYLA SVKAMHEASK KLNECLQEVY EPDWPGRDEA NKIAENNDLL WMDYHQKLVD QALLTMDTYL GQFPDIKSRI AKRGRKLVDY DSARHHYESL QTAKKKDEAK IAKPVSLLEK AAPQWCQGKL QAHLVAQTNL LRNQAEEELI KAQKVFEEMN VDLQEELPSL WNSRVGFYVN TFQSIAGLEE NFHKEMSKLN QNLNDVLVGL EKQHGSNTFT
BIN1 Monoclonal antibody specifically detects BIN1 in Human, Rat samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| BIN1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| ALP-1; AMPH 2; Amph2; amphiphysin 2; amphiphysin II; amphiphysin IIamph2; amphiphysin-like; amphiphysin-like protein; AMPHL; Bin1; box dependant MYC interacting protein 1; box-dependent myc-interacting protein 1; BRAMP-2; bridging integrator 1; DKFZp547F068; MGC10367; MGC105358; myc box dependent interacting protein 1; myc box-dependent-interacting protein 1; SH3 domain-containing protein 9; SH3P9; similar to bridging integrator 1 isoform 9 | |
| Recombinant human BIN1 (immunogen range is 11-280). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| BIN1 |
| K70035_11G6 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| O00499, O08839 | |
| 117028, 274 | |
| IgG1 | |
| Protein A | |
| Antibody |