missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bile Salts, Spectrum™ Chemical
BI109
$895.21 - $12553.42
Specifications
| CAS Max % | 100% |
|---|---|
| Packaging | Poly Bottle |
| Quantity | 100 g |
| Grade | Reagent |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18600980
|
Spectrum Chemical Mfg Cor
BI109100GM |
100 g | Poly Bottle |
Each for $895.21
|
|
||||
|
18600981
|
Spectrum Chemical Mfg Cor
BI109500GM |
500 g | Poly Pail |
Each for $2,443.30
|
|
||||
|
18600982
|
Spectrum Chemical Mfg Cor
BI1095KGBL |
5 kg | Poly Pail |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Bile Salts are also known as bile acids. They are steroidal acids that a thick yellow fluid made by the liver. It helps digest the fat in your diet. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Specifications
| 100% | |
| Poly Bottle | |
| 100 g | |
| Reagent |