missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bile Salts, Spectrum™ Chemical
Description
Spectrum™ Chemical Bile Salts are also known as bile acids. They are steroidal acids that a thick yellow fluid made by the liver. It helps digest the fat in your diet. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
| CAS Max % | 100% |
| Packaging | Poly Pail |
| Quantity | 5 kg |
| Grade | Reagent |