Learn More
Applied Biosystems™ BigDye™ Terminator v1.1 & v3.1 5X Sequencing Buffer
Description
- Optimized for use with BigDye™ Terminator Cycle Sequencing Kits
Order Info
Each
Specifications
Specifications
| Buffer | Sequencing Buffer |
| Concentration | 5X |
| Content And Storage | Store at 2°C to 8°C. |
| For Use With (Equipment) | 3130xl, 3500, 3500xL Genetic Analyzer; GeneAmp™ 9700;Veriti™ Thermal Cycler; 3730, 3730xl DNA Analyzer |
| For Use With (Application) | Standard Sequencing |
| Product Line | BigDye Terminator, BigDye |
| Quantity | 233 mL |
| Shipping Condition | Room Temperature |
| Form | Liquid |
Frequently Asked Questions (FAQs)
A change in the read length is usually caused by something that either changes the migration of the DNA or the efficiency of the sequencing reaction. Some of the things that can cause this are:
• Buffer: Buffer may be prepared incorrectly or on the system for too long.
• Polymer: Polymer may have been on the system the system too long, used past the expiration date, frozen/crystallized and thawed, or mixed with water or buffer during capillary fill. There may be a mismatch between polymer type selected in software and what is installed on the system.
• Environment: Instrument may be operating outside environmental specifications in the Site Preparation Guide. You may have poor/improper instrument ventilation, or air flow may be blowing directly on the system.
• DNA quality: RNA/protein contamination may be reducing the sequencing reaction efficiency. If working with PCR product, there may be carryover of primers and/or dNTPs from the PCR reaction, or the sequencing reaction cleanup is incomplete. If using a kit that has beads in it for the purification, beads may be blocking the capillary tip. If using BigDye XTerminator Purification, heating the XTerminator reagent can cause loss of smaller products.
• DNA quantity: There may be too much DNA competing for entry into the capillary with labeled product, or excessive amounts of DNA creating a temporary blockage of the capillary.
For more information on other causes of short reads and how to address each issue, please refer to the DNA Sequencing by Capillary Electrophoresis: Applied Biosystems Chemistry Guide: Second Edition (Cat. No. 4305080, Rev. C). The guide can be found by searching the Thermo Fisher Scientific website with the catalog number 4305080.
The BigDye Terminator v1.1 and v3.1 chemistries can be diluted, and a 5X Sequencing Buffer is provided with the kit to enable you to do so. However, dilution of the BigDye Terminator may compromise your data integrity depending on the cycle sequencing protocol you are following and the sequence of the particular DNA you are investigating. TheBigDye Direct Cycle Sequencing Kit and the dGTP BigDye Terminator Ready Reaction Kits v1.0 and v3.0 cannot be diluted.
The composition of the BigDye Terminator 5X buffer v1.1 & v3.1 and BigDye Terminator 5X buffer v1.0 & v3.0 chemistries is different and therefore should only be used with the matching chemistry. Use of the incorrect BigDye Terminator 5X buffer can negatively impact sequencing results.
For Research Use Only. Not for use in diagnostic procedures.