missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ BigDye™ M13 HiRes Primers

Catalog No. 4336939
Change view
Click to view available options
Quantity:
100 Reactions
1000 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4336939 1000 Reactions
4336938 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4336939 Supplier Applied Biosystems™ Supplier No. 4336939

The HI-Res Sequencing Primers are optimized primers designed for use with the BigDye™ Terminator BDT Plus Cycle Sequencing Kit to improve sequencing resolution and data quality, particularly at the 5' (five-prime) end of reads.

The HI-Res Sequencing Primers are optimized primers designed for use with the BigDye™ Terminator BDT Plus Cycle Sequencing Kit to improve sequencing resolution and data quality, particularly at the 5' (five-prime) end of reads. Unlike traditional approaches that require dedicated workflows and alternative polymers, HI-Res primers enable enhanced 5' end resolution within standard sequencing workflows using POP-7™ polymer.

Key Features and Benefits

  • Enhance 5' end resolution within standard sequencing workflows using POP-7™ polymer
  • Eliminate the need for dedicated workflows or alternative polymers such as POP-6™
  • Maintain existing workflows with compatibility across BigDye™ Terminator protocols and CE instruments

Product Details

HI-Res Sequencing Primers are specifically engineered to improve sequencing performance at the 5' end of reads, a region that has traditionally been difficult to resolve with standard primer designs. By optimizing primer characteristics and sequencing signal behavior, these primers enable clearer peak resolution and more reliable base calling in early read regions.

Historically, achieving improved 5' end resolution required modified workflows using alternative polymers such as POP-6™, often introducing additional complexity, time, and verification requirements. HI-Res primers enhance 5' end resolution within standard sequencing workflows using POP-7™ polymer.

These primers are designed to work in conjunction with the BigDye™ Terminator BDT Plus Cycle Sequencing Kit, where the combination of optimized primer design and advanced enzyme chemistry enables improved sequencing performance across a wide range of sample types. This integrated system supports consistent results, even when working with challenging templates such as GC-rich regions or structurally complex sequences.

HI-Res primers can be incorporated into current Sanger sequencing workflows. Their compatibility with standard capillary electrophoresis systems and cleanup methods enables easy adoption while enabling improvements in 5' end resolution.

By enabling improved 5' end resolution within existing workflows, HI-Res primers help laboratories generate higher-quality data, simplify operations, and increase overall sequencing efficiency.

Specifications

Format Tube
Isolation Technology Magnetic Bead
Sequencing Type Genome and DNA Sequencing
For Use With (Equipment) SeqStudio™ Genetic Analyzer, SeqStudio™ Flex Series Genetic Analyzer, 3500/3500xL Genetic Analyzer, 3730/3730xl DNA Analyzer Thermal Cyclers: GeneAmp™ PCR System 9700 Dual Series, Veriti™ Fast 96‐well and 384‐well Thermal Cyclers
For Use With (Application) Sanger Sequencing, AT-Rich Sequencing (>65%), Comparative Sequencing (Germline Mutations - 50:50), GC-Rich Sequencing (>65%), Mixed-Base Detection, Long-Read Sequencing, Resequencing, De Novo Sequencing - High Throughput (cDNA), GT-Rich or Difficult Template Sequencing
Product Line BigDye Terminator
Product Type Sequencing Kit
Quantity 1000 Reactions
Technique Fluorescent Dye Terminator Sequencing
Template Compatibility BAC DNA, Lambda DNA, Genomic DNA (Bacterial), Single Stranded DNA, Rolling Circle Amplified Products, Fosmids, PCR Amplicons (50:50 Heterozygotes), Plasmid DNA (≤15 Kb), PCR Amplicons

For Research Use Only.