missing translation for 'onlineSavingsMsg'
Learn More

Bifonazole, 98%, Thermo Scientific™

Catalog No. AC458620010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458620010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC458620010 Supplier Thermo Scientific Chemicals Supplier No. 458620010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 60628-96-8
Molecular Formula C22H18N2
Molecular Weight (g/mol) 310.39
InChI Key OCAPBUJLXMYKEJ-UHFFFAOYSA-N
Synonym bifonazole, mycospor, trifonazole, bifonazol, bifonazolum, amycor, azolmen, bifonazol inn-spanish, bifonazolum inn-latin, 1-4-biphenylyl phenylmethyl-1h-imidazole
PubChem CID 2378
ChEBI CHEBI:78692
IUPAC Name 1-[phenyl-(4-phenylphenyl)methyl]imidazole
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C(C3=CC=CC=C3)N4C=CN=C4

Specifications

Melting Point 149.0°C
Loss on Drying 0.5% max. (1 g, 105°C, 1 h)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 310.39
Percent Purity 98%
Chemical Name or Material Bifonazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 262-336-6

RUO – Research Use Only