missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bifonazole 98.0+%, TCI America™
$196.36 - $196.36
Chemical Identifiers
| CAS | 60628-96-8 |
|---|---|
| Molecular Formula | C22H18N2 |
| Molecular Weight (g/mol) | 310.4 |
| MDL Number | MFCD00865567 |
| InChI Key | OCAPBUJLXMYKEJ-UHFFFAOYSA-N |
| Synonym | bifonazole, mycospor, trifonazole, bifonazol, bifonazolum, amycor, azolmen, bifonazol inn-spanish, bifonazolum inn-latin, 1-4-biphenylyl phenylmethyl-1h-imidazole |
| PubChem CID | 2378 |
| ChEBI | CHEBI:78692 |
| IUPAC Name | 1-[phenyl-(4-phenylphenyl)methyl]imidazole |
| SMILES | C1=CC=C(C=C1)C2=CC=C(C=C2)C(C3=CC=CC=C3)N4C=CN=C4 |
Chemical Identifiers
| 60628-96-8 | |
| 310.4 | |
| OCAPBUJLXMYKEJ-UHFFFAOYSA-N | |
| 2378 | |
| 1-[phenyl-(4-phenylphenyl)methyl]imidazole |
| C22H18N2 | |
| MFCD00865567 | |
| bifonazole, mycospor, trifonazole, bifonazol, bifonazolum, amycor, azolmen, bifonazol inn-spanish, bifonazolum inn-latin, 1-4-biphenylyl phenylmethyl-1h-imidazole | |
| CHEBI:78692 | |
| C1=CC=C(C=C1)C2=CC=C(C=C2)C(C3=CC=CC=C3)N4C=CN=C4 |
Specifications
| 60628-96-8 | |
| White-Yellow | |
| MFCD00865567 | |
| bifonazole, mycospor, trifonazole, bifonazol, bifonazolum, amycor, azolmen, bifonazol inn-spanish, bifonazolum inn-latin, 1-4-biphenylyl phenylmethyl-1h-imidazole | |
| C1=CC=C(C=C1)C2=CC=C(C=C2)C(C3=CC=CC=C3)N4C=CN=C4 | |
| 310.4 | |
| CHEBI:78692 | |
| ≥98.0% (GC,T) | |
| Bifonazole |
| 149°C | |
| C22H18N2 | |
| 5 g | |
| OCAPBUJLXMYKEJ-UHFFFAOYSA-N | |
| 1-[phenyl-(4-phenylphenyl)methyl]imidazole | |
| 2378 | |
| 310.40 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : NI3517000
TSCA : No