missing translation for 'onlineSavingsMsg'
Learn More

Biebrich Scarlet, Water Soluble, Spectrum™ Chemical

$305.82 - $4668.00

Chemical Identifiers

CAS 4196-99-0
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18609068
View Documents
Spectrum Chemical Mfg Cor
BI10525GM
25 g Amber Glass Bottle
Each for $305.82
Only null left
 
18609069
View Documents
Spectrum Chemical Mfg Cor
BI105100GM
100 g Amber Glass Bottle
Each for $613.32
Only null left
 
18609070
View Documents
Spectrum Chemical Mfg Cor
BI1051KGBL
1 kg Amber Glass Bottle
Each for $4,668.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4196-99-0

Specifications

4196-99-0
Reagent
Dark Red
Biebrich Scarlet
1
Amber Glass Bottle
25 g
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.