missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biebrich Scarlet, Water Soluble, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Amber Glass Bottle |
| Color | Dark Red |
| Quantity | 1 kg |
| Grade | Reagent |
| Chemical Name or Material | Biebrich Scarlet |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.